Promega CellTiter-Glo™ Luminescent Cell Viability Assay Kit DFS Item
The most sensitive, fastest, homogeneous cell viability assay. Based on luminescent detection of ATP.
Supplier:  Promega G7572
 View more versions of this product
Catalog No. PR-G7572
Explore available promotions
Includes: CellTiter-Glo Buffer; CellTiter-Glo Substrate (lyophilized)
Description
Description
  • A Luminescent Assay to Determine the Number of Viable Cells in Culture
  • A homogeneous add-mix-measure format reduces the number of steps
  • Higher level of detection requires fewer cells per assay
  • Process plates consecutively with a very stable luminescent signal
  • U.S. Pat. Nos. 6,602,677 and 7,241,584 and other patents and patents pending.
  • U.S. Pat. No. 7,770,310 and other patents and patents pending.
  • U.S. Pat. Nos. 7,083,911, 7,452,663 and 7,732,128 and other patents and patents pending.
  • U.S. Pat. No. 7,741,067 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
Determining the number of viable cells in culture based on the amount of ATP present
96- or 384-well Plates
1000 x 100 μL assays or 4000 x 25 μL assays
100 mL
-30°C to -10°C