Learn More
FuGENE™ 6 Transfection Reagent
FuGENE 6 Transfection Reagent is a nonliposomal formulation designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity.
$463.80 - $3,391.51
Specifications
| Content And Storage | 2°C to 10°C |
|---|---|
| For Use With (Application) | Designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity |
Description
- Reliable Transfection for Common Cell Lines
- Preserve cell viability
- Excellent compatibility with everyday cell lines
- Straightforward protocol; minimal optimization
Specifications
| 2°C to 10°C | |
| Designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity |
FuGENE™ HD is sold with a Limited Use Label License (LULL). You shall use this Product in accordance with all applicable laws, rules and regulations. You may not attempt to reverse engineer this Product. If you are not willing to accept these conditions, the distributor from whom the Product was purchased will accept the return of the unopened Product and provide you with a full refund. However, in the event this Product is opened, you agree to be bound by the conditions of this limited use statement.