Promega FuGENE™ 6 Transfection Reagent DFS Item
FuGENE 6 Transfection Reagent is a nonliposomal formulation designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity.
Supplier:  Promega E2692
 View more versions of this product
Catalog No. PRE2692
Explore available promotions
Description
Description
  • Reliable Transfection for Common Cell Lines
  • Preserve cell viability
  • Excellent compatibility with everyday cell lines
  • Straightforward protocol; minimal optimization
Specifications
Specifications
2°C to 10°C
Designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity
5 x 1 mL