missing translation for 'onlineSavingsMsg'
Learn More

Promega Go Taq™ Master Mixes DFS Item

New formulation of Taq DNA Polymerase with enhanced performance over standard Taq DNA Polymerase. Available in two all-in-one master mixes: green and colorless. The green master mix allows direct gel analysis after amplification.

$72.47 - $625.87

Specifications

Concentration 2X
For Use With (Application) Standard PCR
View More Specs

Includes: GoTaq Green Master Mix or GoTaq Colorless Master Mix, Nuclease-free water

Products 4
Catalog Number Mfr. No. Description No. of Reactions Price Quantity  
Catalog Number Mfr. No. Description No. of Reactions Price Quantity  
PRM7133 DFS Item
View Documents
Promega
M7133
Go Taq™ Colorless Master Mix 1000 Reactions N/A
Only null left
 
PRM7132 DFS Item
View Documents
Promega
M7132
Go Taq™ Colorless Master Mix 100 Reactions N/A
Only null left
 
PR-M7122 DFS Item
View Documents
Promega
M7122
Go Taq™ Green Master Mix 100 Reactions N/A
Only null left
 
PR-M7123 DFS Item
View Documents
Promega
M7123
Go Taq™ Green Master Mix 1000 Reactions N/A
Only null left
 
Description

Description

  • All-in-One Optimized PCR Master Mix-Just Add Template, Primers and Water
  • Directly substitute into your current PCR conditions
  • Use the green master mix for direct-to-gel analysis after amplification
  • Use the colorless master mix for post-amplification analysis by fluorescence or absorbance without prior DNA purification

  • The purchase of this product conveys to the buyer the limited, nonexclusive, nontransferable right (without the right to resell, repackage, or further sublicense) under U.S. Published Patent Appln. 20020150900 and U.S. Pat. Nos. 5,432,272, 6,617,106 and 6,140,496 to use the product. No other license is granted to the buyer whether expressly, by implication, by estoppel or otherwise. In particular, the purchase of this product does not include or carry any right or license to sell this product. For information on purchasing a license for other uses, please contact Promega Corporation, Licensing, 2800 Woods Hollow Road, Madison, WI 53711, or EraGen Biosciences, Corporate Licensing, 918 Deming Way, Suite 201, Madison, WI 53717. Phone (608) 662-9000; Fax (608) 662-9003.
  • This product is designed and sold for use in the multiplex PCR process covered by U.S. Pat. No. 5,582,989 and Canadian Pat. No. 1,339,731. A limited license has been granted under the patent to use only this amount of the product to practice the multiplex PCR process and is conveyed to the purchaser by the purchase of this product.

Specifications

Specifications

2X
Standard PCR
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'