missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega GoTaq™ Probe qPCR Master Mix
GoTaq™ Probe qPCR Master Mix is a ready-to-use stabilized 2X formulation containing GoTaq™ Hot Start Polymerase, MgCl2, dNTPs and proprietary reaction buffer.
$797.15 - $797.15
Specifications
For Use With (Application) | Gene Expression |
---|---|
Compatibility | Both standard and fast instrument cycling programs |
Concentration | 2X |
Catalog Number | Mfr. No. | No. of Reactions | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Quantity | Price | Quantity | ||||
PRA6102
|
Promega
A6102 |
1000 Reactions | 10 mL |
N/A
|
|
||||
Description
- Convenient qPCR Amplification Using Hydrolysis Detection Probes
- Ready-to-use mix of optimized components for robust amplification
- Compatible with both fast and standard cycling methods
- Sensitive detection for quantitation over a broad dynamic range
Specifications
Gene Expression | |
Both standard and fast instrument cycling programs | |
2X |