missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega GoTaq™ Probe qPCR Master Mix DFS Item

GoTaq™ Probe qPCR Master Mix is a ready-to-use stabilized 2X formulation containing GoTaq™ Hot Start Polymerase, MgCl2, dNTPs and proprietary reaction buffer.

Supplier:  Promega A6102

 View more versions of this product

Catalog No. PRA6102

Explore available promotions

Description

Description

  • Convenient qPCR Amplification Using Hydrolysis Detection Probes
  • Ready-to-use mix of optimized components for robust amplification
  • Compatible with both fast and standard cycling methods
  • Sensitive detection for quantitation over a broad dynamic range
Specifications

Specifications

• 2X GoTaq Probe qPCR Master Mix (dTTP), 10 x 1 mL
• 30μM CXR Reference Dye, 2 x 200 μL
• Nuclease-free Water, 13 mL

Store at –30° to –10°C.
Gene Expression
2X
10 mL
Both standard and fast instrument cycling programs
1000 Reactions
Videos
SDS
Documents

Documents

Promotions

Promotions