missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega GSH/GSSG-Glo™ Glutathione Assay
The GSH/GSSG-Glo™ Assay is a luminescence-based system for the detection and quantification of total glutathione (GSH +GSSG), GSSG and GSH/GSSG ratios in cultured cells.
$1047.64 - $4373.52
Specifications
Content And Storage | -30°C to -10°C |
---|---|
For Use With (Application) | For the detection and quantification of total Glutathione (GSH +GSSG), GSSG and GSH/GSSG ratios in cultured cells |
For Use With (Equipment) | For rapid multiwell plate |
Catalog Number | Mfr. No. | Kit Contents | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Kit Contents | Quantity | Price | Quantity | ||||
PRV6611
|
Promega
V6611 |
1 x 250μL DTT 100mM, 1 x 100μL Luciferin-NT, 1 x 500μL Glutathione S-transferase solution, 1 x 100μL Glutathione 5mM, 1 x 100μL Luciferin detection reagent, 1 x 10mL | 10 mL |
Each for $1,047.64
|
|
||||
PRV6612
|
Promega
V6612 |
1 x 1.25mL DTT 100mM, 1 x 500μL Luciferin-NT, 3 x 500μL Glutathione S-transferase solution, 1 x 500μL Glutathione 5mM, 1 x 1 each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer with Estera | 50 mL |
Each for $4,373.52
|
|
||||
Description
- A Luminescent Assay for the Detection and Quantification of Total Glutathione Ratios
- Measures physiologically relevant GSH/GSSG ratios
- Simple protocol minimizes sample handling, reducing variability
Specifications
-30°C to -10°C | |
For the detection and quantification of total Glutathione (GSH +GSSG), GSSG and GSH/GSSG ratios in cultured cells | |
For rapid multiwell plate |