Learn More
Promega GSH/GSSG-Glo™ Glutathione Assay DFS Item
The GSH/GSSG-Glo™ Assay is a luminescence-based system for the detection and quantification of total glutathione (GSH +GSSG), GSSG and GSH/GSSG ratios in cultured cells.
Supplier:  Promega V6611
 View more versions of this product
Catalog No. PRV6611
Only null left
Explore available promotions
Description
Description
  • A Luminescent Assay for the Detection and Quantification of Total Glutathione Ratios
  • Measures physiologically relevant GSH/GSSG ratios
  • Simple protocol minimizes sample handling, reducing variability
Specifications
Specifications
10 mL
For the detection and quantification of total Glutathione (GSH +GSSG), GSSG and GSH/GSSG ratios in cultured cells
For rapid multiwell plate
-30°C to -10°C
1 x 250μL DTT 100mM, 1 x 100μL Luciferin-NT, 1 x 500μL Glutathione S-transferase solution, 1 x 100μL Glutathione 5mM, 1 x 100μL Luciferin detection reagent, 1 x 10mL
Product Suggestions
Product Suggestions