missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Renilla-Glo™ Luciferase Assay System
The Renilla-Glo™ Luciferase Assay System is a single-addition reagent that generates a glow-type signal with Renilla luciferase.
$324.13 - $14541.04
Specifications
Content And Storage | -30°C to -10°C |
---|
Includes: Renilla-Glo Luciferse Assay Substrate and Renilla-Glo Luciferase Assay Buffer
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PRE2710
|
Promega
E2710 |
10 mL |
Each for $324.13
|
|
|||||
PRE2750
|
Promega
E2750 |
10 x 100 mL |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PRE2720
|
Promega
E2720 |
100 mL |
Each for $1,727.37
|
|
|||||
Description
- Simple Add-and-Read Renilla Luciferase Reporter Assay
- No need for separate lysis and reagent injection steps
- Extended bright light output optimized for batch and continuous-processing
- Low background provides increased sensitivity
Specifications
-30°C to -10°C |
- U.S. Patents 7,078,181; 7,108,996; 7,118,878; Australian Patent 2001275325 and other patents pending.
- This product does not convey a license to use recombinant Renilla luciferase under U.S. Patents 5,292,658; 5,418,155 and related patents. Promega sells licensed Renilla luciferase vectors, which may be used in conjunction with this product.
- Certain applications of this product may require licenses from others.