Promega Renilla-Glo™ Luciferase Assay System DFS Item
The Renilla-Glo™ Luciferase Assay System is a single-addition reagent that generates a glow-type signal with Renilla luciferase.
Supplier:  Promega E2720
 View more versions of this product
Catalog No. PRE2720
Explore available promotions
Includes: Renilla-Glo Luciferse Assay Substrate and Renilla-Glo Luciferase Assay Buffer
Description
Description
  • Simple Add-and-Read Renilla Luciferase Reporter Assay
  • No need for separate lysis and reagent injection steps
  • Extended bright light output optimized for batch and continuous-processing
  • Low background provides increased sensitivity
Specifications
Specifications
-30°C to -10°C
100 mL
Videos
SDS
Documents
Documents
Promotions
Promotions
  • U.S. Patents 7,078,181; 7,108,996; 7,118,878; Australian Patent 2001275325 and other patents pending.
  • This product does not convey a license to use recombinant Renilla luciferase under U.S. Patents 5,292,658; 5,418,155 and related patents. Promega sells licensed Renilla luciferase vectors, which may be used in conjunction with this product.
  • Certain applications of this product may require licenses from others.