missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Renilla Luciferase Assay System
Renilla Luciferase Assay System provides a fast, sensitive method for detecting luciferase from sea pansy (Renilla reniformis).
$304.36 - $1529.73
Specifications
Content And Storage | -30°C to -10°C |
---|---|
For Use With (Application) | Detecting the Luciferase from Sea pansy (Renilla reniformis) |
Includes: Renilla Luciferase Assay Substrate, Renilla Luciferase Assay Buffer, Renilla Luciferase Assay Lysis Buffer (5X)
Description
- Fast, Sensitive Detection of Renilla Luciferase Activity
- Linear range extends over 7 logs
- Low background, increased sensitivity
- Detect 10-19 moles of Renilla luciferase
- U.S. Patents 7,078,181; 7,108,996; 7,118,878; Australian Patent 2001275325 and other patents pending.
- Certain applications of this product may require licenses from others.
- This product does not convey a license to use recombinant Renilla luciferase under U.S. Patent 5,418,155 and related patents. Promega sells licensed Renilla luciferase vectors, which may be used in conjunction with this product.
Specifications
-30°C to -10°C | |
Detecting the Luciferase from Sea pansy (Renilla reniformis) |