Learn More
Promega Renilla Luciferase Assay System DFS Item
Renilla Luciferase Assay System provides a fast, sensitive method for detecting luciferase from sea pansy (Renilla reniformis).
Supplier:  Promega E2810
 View more versions of this product
Catalog No. PR-E2810
Only null left
Explore available promotions
Includes: Renilla Luciferase Assay Substrate, Renilla Luciferase Assay Buffer, Renilla Luciferase Assay Lysis Buffer (5X)
Description
Description
  • Fast, Sensitive Detection of Renilla Luciferase Activity
  • Linear range extends over 7 logs
  • Low background, increased sensitivity
  • Detect 10-19 moles of Renilla luciferase
  • U.S. Patents 7,078,181; 7,108,996; 7,118,878; Australian Patent 2001275325 and other patents pending.
  • Certain applications of this product may require licenses from others.
  • This product does not convey a license to use recombinant Renilla luciferase under U.S. Patent 5,418,155 and related patents. Promega sells licensed Renilla luciferase vectors, which may be used in conjunction with this product.
Specifications
Specifications
-30°C to -10°C
Detecting the Luciferase from Sea pansy (Renilla reniformis)
100 Assays
Product Suggestions
Product Suggestions