Learn More
Promega Kinase-Glo™ Plus Luminescent Kinase Assay System
The Kinase-Glo™ Luminescent Kinase Assays are homogeneous non-radioactive systems for determining the activity of purified kinases by quantifying the amount of ATP remaining in solution following a kinase reaction.
$288.56 - $8408.92
Specifications
| Product Type | Kinase-Glo Plus |
|---|---|
| pH | 7 |
| Content And Storage | -20°C |
| For Use With (Application) | Automated high throughput screening and they can be used to assay protein, lipid and sugar kinases |
| Form | Fluid |
Description
- Monitors Activity of Purified Kinases by Quantifying ATP
- Luminescent signal is inversely proportional to the amount of kinase activity
- Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP
Specifications
| Kinase-Glo Plus | |
| -20°C | |
| Fluid |
| 7 | |
| Automated high throughput screening and they can be used to assay protein, lipid and sugar kinases |
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.