Promega Kinase-Glo™ Plus Luminescent Kinase Assay System DFS Item
The Kinase-Glo™ Luminescent Kinase Assays are homogeneous non-radioactive systems for determining the activity of purified kinases by quantifying the amount of ATP remaining in solution following a kinase reaction.
Supplier:  Promega V3772
 View more versions of this product
Catalog No. PR-V3772
Explore available promotions
Description
Description
  • Monitors Activity of Purified Kinases by Quantifying ATP
  • Luminescent signal is inversely proportional to the amount of kinase activity
  • Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP
Specifications
Specifications
Kinase-Glo Plus
-20°C
10 x 10 mL
7
Automated high throughput screening and they can be used to assay protein, lipid and sugar kinases
Fluid
Videos
SDS
Documents
Documents
Promotions
Promotions
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.