Promega ONE-Glo™ Assay DFS Item
The ONE-Glo™ + Tox Assay combines luciferase assay chemistry with a cell viability marker to better understand reporter gene expression in the context of cell health. The assay uses a two-step, addition-only process to measure in a single well of a plate.
Supplier:  Promega E7110
 View more versions of this product
Catalog No. PRE7110
Explore available promotions
Description
Description
  • Sensitive Detection of Reporter Activity and Cell Health
  • Measures cell viability and firefly luciferase activity in the same assay well
  • Easy to use—simply add reagent, mix and read
  • Scalable to meet throughput needs, up to 1,536-well format
Specifications
Specifications
1 Plate
1 x 10mL ONE-Glo™ Luciferase Assay Buffer, 1 x 1 Vial ONE-Glo™ Luciferase assay substrate (lyophilized), 1 x 10μL GF-AFC substrate, Assay Buffer 1 x 10mL
Fluid
-30°C to -10°C
7.5
Product Suggestions
Product Suggestions