Promega pGEM™-T and pGEM™-T Easy Vector Systems DFS Item
Convenient systems for the routine subcloning of PCR products
Supplier:  Promega A3600
 View more versions of this product
Catalog No. PR-A3600
Explore available promotions
Includes: Vector, Control insert DNA, T4 DNA ligase, Ligation buffer, High efficiency competent cells (Mfr. Nos. A3610 and A1380 only)
Description
Description
The pGEM-T Vector is prepared by cutting Promega's pGEM-5Zf(+) Vector with EcoR V and adding a 3« terminal thymidine to both ends. These single 3«-T overhangs at the insertion site greatly improve the efficiency of ligation of a PCR product into the plasmid.
  • 2X Rapid Ligation Buffer allows reactions to be completed in one hour at room temperature Blue/white screening can be used to directly identify recombinant clones, as T7 and SP6 RNA polymerase promoters flank a multiple cloning region within the α-peptide coding region for β-galactosidase, permitting insertional inactivation of the α-peptide
  • f1 Origin of Replication allows the preparation of single-stranded DNA
pGEM-T Vector Systems
  • Multiple cloning site is flanked by recognition sites for the restriction enzyme BstZ I Single-enzyme digestion allows release of the insert
  • Double digestion may also be used to release the insert from the vector
pGEM-T Easy Vector Systems
  • Recognition sites for BstZ I, EcoR I, and Not I flank the insertion site Several options are available for removal of the desired insert DNA with a single restriction digestion
Specifications
Specifications
SP6, T7
-30°C to -10°C
pGEM-T vector I
BstZI
20 Reactions
For the routine subcloning of PCR products