missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega PureYield™ Plasmid Maxiprep System
The PureYield Plasmid Maxiprep System isolates transfection-quality plasmid DNA. Yields up to 1mg of plasmid DNA from a 250ml bacterial culture.
$315.93 - $1741.31
Specifications
Content And Storage | 15°C to 30°C |
---|---|
Format | Vacuum |
For Use With (Equipment) | Vac-Man™ laboratory vacuum manifold |
For Use With (Application) | Isolates transfection-quality plasmid DNA |
Sample Volume | 250mL |
Includes: All systems include reagents and columns for purification. A6730 also includes vacuum pump, vacuum manifold, and vacuum elution device.
Catalog Number | Mfr. No. | Includes | No. of Reactions | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | No. of Reactions | Price | Quantity | ||||
PRA2392
|
Promega
A2392 |
130mL Neutralization solution, 2 × 5 PureYield clearing columns, 2 × 5 PureYield maxi binding columns, 36.5mL Endotoxin removal wash, 82mL Column wash, 125mL Cell resuspension solution, 125mL Cell lysis solution, 25mL Nuclease-free water | 10 Preps |
Each for $315.93
|
|
||||
PRA2393
|
Promega
A2393 |
315mL Neutralization solution, 5 × 5 PureYield clearing columns, 5 × 5 PureYield maxi binding columns, 85.3mL Endotoxin removal wash, 210mL Column wash, 315mL Cell resuspension solution, 315mL Cell lysis solution, 2 × 25mL Nuclease-free | 25 Preps |
Each for $732.73
|
|
||||
PRA6730
|
Promega
A6730 |
PureYield plasmid maxiprep system (2 × 25 preps), 4 Eluator vacuum elution devices, Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold | 2 x 25 preps |
Each for $1,741.31
|
|
||||
Description
- Up to 1mg of Transfection-Ready Plasmid from 250mL Cultures
- One-hour, vacuum-based protocol
- Highly pure, concentrated plasmid DNA
- No tedious, gravity-drip columns or post-elution alcohol precipitation
Specifications
15°C to 30°C | |
Vac-Man™ laboratory vacuum manifold | |
250mL |
Vacuum | |
Isolates transfection-quality plasmid DNA | |
Up to 1mg |