missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega PureYield™ Plasmid Maxiprep System
The PureYield Plasmid Maxiprep System isolates transfection-quality plasmid DNA. Yields up to 1mg of plasmid DNA from a 250ml bacterial culture.
Supplier: Promega A6730
Includes: All systems include reagents and columns for purification. A6730 also includes vacuum pump, vacuum manifold, and vacuum elution device.
Description
- Up to 1mg of Transfection-Ready Plasmid from 250mL Cultures
- One-hour, vacuum-based protocol
- Highly pure, concentrated plasmid DNA
- No tedious, gravity-drip columns or post-elution alcohol precipitation
Specifications
15°C to 30°C | |
Vac-Man™ laboratory vacuum manifold | |
PureYield plasmid maxiprep system (2 × 25 preps), 4 Eluator vacuum elution devices, Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold | |
Plasmid Maxiprep Start-Up Kit | |
250mL | |
Up to 1mg |
Vacuum | |
Isolates transfection-quality plasmid DNA | |
2 x 25 preps | |
1/Ea. | |
110V |