Promega ReliaPrep™ RNA Miniprep System DFS Item
The ReliaPrep RNA Miniprep Systems provide a fast and simple technique for preparation of intact total RNA from cultured cells or tissue in as little as 30 minutes from very small amounts of input material.
Supplier:  Promega Z6012
 View more versions of this product
Catalog No. PRZ6012
Explore available promotions
Includes: ReliaPrep Minicolumns, Collection Tubes, 1-Thioglycerol, Column Wash Solution (CWE), Nuclease-Free Water, BL Buffer, Elution Tubes, RNA Wash Solution (RWA), Yellow Core Buffer, 0.09M MnCl2, DNase I (lyophilized)
Description
Description
  • Efficiently Purify RNA from as Few as 100 Cells or 0.25mg Tissue
  • Simple, 30-minute protocol
  • Elutes in small volume, generating highly concentrated RNA
  • No phenol/chloroform or ß-mercaptoethanol required
Specifications
Specifications
15°C to 30°C
Collection tubes 2 x 250, ReliaPrep™ Minicolumns 5 x 50, 1-Thioglycerol 2 x 900μL, column wash solution (CWE) 1 x 24mL, Nuclease-free Water 1 x 13mL, BL Buffer 1 x 150mL, elution tubes 5 x 50, RNA wash solution (RWA) 1
250 Preps
Purify RNA from cultured cells
RNA Kit