Promega ReliaPrep™ RNA Miniprep System DFS Item
The ReliaPrep RNA Miniprep Systems provide a fast and simple technique for preparation of intact total RNA from cultured cells or tissue in as little as 30 minutes from very small amounts of input material.
$185.57 - $2330.88
Specifications
Content And Storage 15°C to 30°C
Product Type RNA Kit
View More Specs
Includes: ReliaPrep Minicolumns, Collection Tubes, 1-Thioglycerol, Column Wash Solution (CWE), Nuclease-Free Water, BL Buffer, Elution Tubes, RNA Wash Solution (RWA), Yellow Core Buffer, 0.09M MnCl2, DNase I (lyophilized)
Products 5
Catalog Number Mfr. No. For Use With (Application) Includes Quantity Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Includes Quantity Price Quantity  
PRZ6010 DFS Item
View Documents
Promega
Z6010
Purify RNA from cultured cells Collection tubes 2 x 10, ReliaPrep™ Minicolumns 1 x 10, 1-Thioglycerol 1 x 75μL, column wash solution (CWE) 1 x 1mL, Nuclease-free Water 1 x 1,250μL, BL Buffer 2 x 3.25mL, elution tubes 1 x 25, RNA was 10 Preps
Each for $185.57
PRZ6012 DFS Item
View Documents
Promega
Z6012
Purify RNA from cultured cells Collection tubes 2 x 250, ReliaPrep™ Minicolumns 5 x 50, 1-Thioglycerol 2 x 900μL, column wash solution (CWE) 1 x 24mL, Nuclease-free Water 1 x 13mL, BL Buffer 1 x 150mL, elution tubes 5 x 50, RNA wash solution (RWA) 1 250 Preps
Each for $2,297.97
PRZ6110 DFS Item
View Documents
Promega
Z6110
Purify RNA from tissues Collection tubes 2 x 10, ReliaPrep™ Minicolumns 1 x 10, 1-Thioglycerol 2 x 75μL, column wash solution (CWE) 1 x 1mL, Nuclease-free Water 1 x 1250mL, Lysis Buffer (LBA) 1 x 6mL, elution tubes 1 x 25, RNA wash solution (R 10 Preps
Each for $201.59
PRZ6112 DFS Item
View Documents
Promega
Z6112
Purify RNA from tissues Collection tubes 2 x 250, ReliaPrep™ Minicolumns 5 x 50, 1-Thioglycerol 4 x 900μL, column wash solution (CWE) 1 x 24mL, Nuclease-free Water 1 x 13mL, Lysis Buffer (LBA) 1 x 150mL, elution tubes 5 x 50, RNA wash solution 250 Preps
Each for $2,330.88
PRZ6111 DFS Item
View Documents
Promega
Z6111
Purify RNA from tissues Collection tubes 2 x 50, ReliaPrep™ Minicolumns 1 x 50, 1-Thioglycerol 2 x 900μL, column wash solution (CWE) 1 x 5mL, Nuclease-free Water 1 x 13mL, Lysis Buffer (LBA) 1 x 30mL, elution tubes 1 x 50, RNA wash solution (R 50 Preps
Each for $509.35
Description
Description
  • Efficiently Purify RNA from as Few as 100 Cells or 0.25mg Tissue
  • Simple, 30-minute protocol
  • Elutes in small volume, generating highly concentrated RNA
  • No phenol/chloroform or ß-mercaptoethanol required
Specifications
Specifications
15°C to 30°C
RNA Kit
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'