Promega ReliaPrep™ RNA Miniprep System
The ReliaPrep RNA Miniprep Systems provide a fast and simple technique for preparation of intact total RNA from cultured cells or tissue in as little as 30 minutes from very small amounts of input material.
Supplier: Promega Z6111
Includes: ReliaPrep Minicolumns, Collection Tubes, 1-Thioglycerol, Column Wash Solution (CWE), Nuclease-Free Water, BL Buffer, Elution Tubes, RNA Wash Solution (RWA), Yellow Core Buffer, 0.09M MnCl2, DNase I (lyophilized)
Description
- Efficiently Purify RNA from as Few as 100 Cells or 0.25mg Tissue
- Simple, 30-minute protocol
- Elutes in small volume, generating highly concentrated RNA
- No phenol/chloroform or Ã-mercaptoethanol required
Specifications
15°C to 30°C | |
Collection tubes 2 x 50, ReliaPrep™ Minicolumns 1 x 50, 1-Thioglycerol 2 x 900μL, column wash solution (CWE) 1 x 5mL, Nuclease-free Water 1 x 13mL, Lysis Buffer (LBA) 1 x 30mL, elution tubes 1 x 50, RNA wash solution (R | |
50 Preps |
Purify RNA from tissues | |
RNA Kit |