missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega RNase One™
Single-strand-specific RNA endonuclease. Cleaves between all ribonucleotides.
$137.99 - $466.28
Specifications
Concentration | 5 to 10 U/μL |
---|---|
Color | Colorless |
Components | 10X Reaction Buffer: Tris HCl 100mM (pH 7.5 at 25°C), EDTA 50mM, Sodium Acetate 2M |
pH | 7.3 |
For Use With (Application) | Mapping, RNase protection assay, mismatch detection, selective removal of RNA from DNA preparations |
Description
- Degrades RNA to Cyclic Nucleotide Monophosphate (NMP) Intermediates
- Ability to cleave a phosphodiester bond between any two ribonucleotides
- Supplied at a concentration of 5-10u/μL
Specifications
5 to 10 U/μL | |
10X Reaction Buffer: Tris HCl 100mM (pH 7.5 at 25°C), EDTA 50mM, Sodium Acetate 2M | |
Mapping, RNase protection assay, mismatch detection, selective removal of RNA from DNA preparations | |
-30°C to -10°C | |
Ribonuclease | |
Liquid |
Colorless | |
7.3 | |
10mM Tris HCl (pH 8), 200mM NaCl, 50% Glycerol | |
Recombinant E. coli strain | |
27 |