Promega RNase One™ DFS Item
Single-strand-specific RNA endonuclease. Cleaves between all ribonucleotides.
Supplier:  Promega M4261
 View more versions of this product
Catalog No. PR-M4261
Explore available promotions
Description
Description
  • Degrades RNA to Cyclic Nucleotide Monophosphate (NMP) Intermediates
  • Ability to cleave a phosphodiester bond between any two ribonucleotides
  • Supplied at a concentration of 5-10u/μL
Specifications
Specifications
5 to 10 U/μL
10X Reaction Buffer: Tris HCl 100mM (pH 7.5 at 25°C), EDTA 50mM, Sodium Acetate 2M
Mapping, RNase protection assay, mismatch detection, selective removal of RNA from DNA preparations
-30°C to -10°C
1000 U
27
Colorless
7.3
10mM Tris HCl (pH 8), 200mM NaCl, 50% Glycerol
Recombinant E. coli strain
Ribonuclease
Liquid