missing translation for 'onlineSavingsMsg'
Learn More

Promega SV Total RNA Isolation System DFS Item

A fast, simple technique for preparation of intact total RNA from tissues, cultured cells and white blood cells in as little as one hour.

$347.84 - $4965.88

Specifications

Content And Storage 15°C to 30°C
Format Spin Column
For Use With (Application) Molecular biology applications, including RT-PCR and northern blotting
View More Specs

Includes: SV Total RNA Isolation System: Spin column assemblies and elution tubes, DNase I (lyophilized), SV RNA lysis buffer, SV RNA dilution buffer, β-mercaptoethanol, MnCl2 (0.09M), Yellow core buffer, SV DNase stop solution, SV RNA wash solution, Nuclease-free water
SV Total RNA Isolation Start-Up Kit (110V): SV Total RNA Isolation System, Vacuum Adapters, Welch™ Vacuum Pump for North America Electrical, Vac-Man™ Laboratory Vacuum Manifold (20-sample capacity)

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRZ3105 DFS Item
View Documents
Promega
Z3105
250 Preps
Each for $2,727.43
Only null left
 
PR-Z3101 DFS Item
View Documents
Promega
Z3101
10 Preps
Each for $347.84
Only null left
 
PRZ3380 DFS Item
View Documents
Promega
Z3380
1/Ea.
Each for $4,965.88
Only null left
 
PR-Z3100 DFS Item
View Documents
Promega
Z3100
50 Preps
Each for $594.24
Only null left
 
Description

Description

  • Extracts RNA from Tissues, Cultured Cells and White Blood Cells
  • Rapid isolation of high yields of total RNA without the use of hazardous compounds
  • Choice of microcentrifugation (spin) or vacuum purification methods
  • Purified RNA suitable for all routine molecular biology applications, including RT-PCR
Specifications

Specifications

15°C to 30°C
Spin Column
Molecular biology applications, including RT-PCR and northern blotting
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'