Promega SV Total RNA Isolation System DFS Item
A fast, simple technique for preparation of intact total RNA from tissues, cultured cells and white blood cells in as little as one hour.
Supplier:  Promega Z3380
 View more versions of this product
Catalog No. PRZ3380
Explore available promotions
Includes:
  • SV Total RNA Isolation System: Spin column assemblies and elution tubes, DNase I (lyophilized), SV RNA lysis buffer, SV RNA dilution buffer, β-mercaptoethanol, MnCl2 (0.09M), Yellow core buffer, SV DNase stop solution, SV RNA wash solution, Nuclease-free water
  • SV Total RNA Isolation Start-Up Kit (110V): SV Total RNA Isolation System, Vacuum Adapters, Welch™ Vacuum Pump for North America Electrical, Vac-Man™ Laboratory Vacuum Manifold (20-sample capacity)
Description
Description
  • Extracts RNA from Tissues, Cultured Cells and White Blood Cells
  • Rapid isolation of high yields of total RNA without the use of hazardous compounds
  • Choice of microcentrifugation (spin) or vacuum purification methods
  • Purified RNA suitable for all routine molecular biology applications, including RT-PCR
Specifications
Specifications
15°C to 30°C
Molecular biology applications, including RT-PCR and northern blotting
1/Ea.
Spin Column
Spin column assemblies and elution tubes, DNase I (lyophilized), SV RNA Lysis Buffer, SV RNA dilution Buffer, beta-mercaptoethanol, MnCl2 (0.09M), yellow core Buffer, SV DNase stop solution, SV RNA wash solution, Nuclease-free Water