Promega TNT™ T7 Insect Cell Extract Protein Expression System DFS Item
The TNT T7 Insect Cell Extract Protein Expression System is a convenient, quick, single-tube, coupled transcription/translation system for cell-free protein expression.
Supplier:  Promega L1101
 View more versions of this product
Catalog No. PRL1101
Explore available promotions
Includes: TNT T7 ICE Master Mix, Luciferase ICE T7 Control DNA, Nuclease-free water
Description
Description
  • Single-Tube, Coupled Transcription/Translation for Cell-Free Protein Expression using Insect Cell Extract
  • Quick, single-tube, coupled transcription/translation system for the cell-free expression of proteins from DNA templates
  • Extract derived from Spodoptera frugiperda Sf21 cell line
  • Proteins expressed from genes cloned downstream of the T7 promoter; companion vectors available
Specifications
Specifications
TnT™ T7 ICE master mix, luciferase ICE T7 control DNA, Nuclease-free Water
75 μg/mL
10 Reactions
<-65°C
4 hr.
Insect Cell Protein Expression System
10 reactions