Promega TNT™ T7 Insect Cell Extract Protein Expression System DFS Item
The TNT T7 Insect Cell Extract Protein Expression System is a convenient, quick, single-tube, coupled transcription/translation system for cell-free protein expression.
$417.68 - $1446.72
Specifications
Kit Contents TnT™ T7 ICE master mix, luciferase ICE T7 control DNA, Nuclease-free Water
Test Time 4 hr.
Yield 75 μg/mL
Type Insect Cell Protein Expression System
Content And Storage <-65°C
View More Specs
Includes: TNT T7 ICE Master Mix, Luciferase ICE T7 Control DNA, Nuclease-free water
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRL1101 DFS Item
View Documents
Promega
L1101
10 reactions
Each for $417.68
PRL1102 DFS Item
View Documents
Promega
L1102
40 reactions
Each for $1,446.72
Description
Description
  • Single-Tube, Coupled Transcription/Translation for Cell-Free Protein Expression using Insect Cell Extract
  • Quick, single-tube, coupled transcription/translation system for the cell-free expression of proteins from DNA templates
  • Extract derived from Spodoptera frugiperda Sf21 cell line
  • Proteins expressed from genes cloned downstream of the T7 promoter; companion vectors available
Specifications
Specifications
TnT™ T7 ICE master mix, luciferase ICE T7 control DNA, Nuclease-free Water
75 μg/mL
<-65°C
4 hr.
Insect Cell Protein Expression System
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'