Promega TNT™ T7 Insect Cell Extract Protein Expression System
The TNT T7 Insect Cell Extract Protein Expression System is a convenient, quick, single-tube, coupled transcription/translation system for cell-free protein expression.
Supplier: Promega L1102
Includes: TNT T7 ICE Master Mix, Luciferase ICE T7 Control DNA, Nuclease-free water
Description
- Single-Tube, Coupled Transcription/Translation for Cell-Free Protein Expression using Insect Cell Extract
- Quick, single-tube, coupled transcription/translation system for the cell-free expression of proteins from DNA templates
- Extract derived from Spodoptera frugiperda Sf21 cell line
- Proteins expressed from genes cloned downstream of the T7 promoter; companion vectors available
Specifications
TnT™ T7 ICE master mix, luciferase ICE T7 control DNA, Nuclease-free Water | |
75 μg/mL | |
40 Reactions | |
<-65°C |
4 hr. | |
Insect Cell Protein Expression System | |
40 reactions |