missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega TNT™ T7 Insect Cell Extract Protein Expression System Companion Vectors DFS Item

Designed for use with Insect Cell Extracts and contain untranslated region (UTR) sequences at the 5' and 3' ends of the gene coding region to enhance translation efficiency.

$565.25 - $565.25

Specifications

Promoter T7
Restriction Site SgfI and PmeI
Storage Buffer 10mM Tris HCl, 1mM EDTA (pH 8)
Content And Storage -30°C to -10°C
Concentration 100 ng/μL
View More Specs

Products 2
Catalog Number Mfr. No. Vector Price Quantity  
Catalog Number Mfr. No. Vector Price Quantity  
PRL1081 DFS Item
missing translation for 'viewDocuments'
Promega
L1081
pF25K ICE T7 Flexi™ vector
Each for $565.25
 
PRL1061 DFS Item
missing translation for 'viewDocuments'
Promega
L1061
pF25A ICE T7 Flexi™ vector
Each for $565.25
 
Description

Description

  • Protein Expression Vectors Used With Insect Cell Extracts for In Vitro Translation
  • Efficient transfer of Flexi(R) vector inserts for direct use of recombinant clones, minimizing time wasted screening background colonies
  • Adaptable to high-throughput formats for large screening projects
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or mammalian protein expression) as needed
Specifications

Specifications

T7
10mM Tris HCl, 1mM EDTA (pH 8)
100 ng/μL
Clone ORF and conveniently transfer these inserts to a variety of vectors designed for multiple applications
SgfI and PmeI
-30°C to -10°C
20 μg
Videos
SDS
Documents

Documents

  • Patent Pending.
  • For research use only. Persons wishing to use this product or its derivatives in other fields of use, including without limitation, commercial sale, diagnostics or therapeutics, should contact Promega Corporation for licensing information.
  • Use of this vector for applications outside of the TNT T7 Insect Cell Extract Protein Expression System may require a license from Shimadzu Corporation. For more information contact: t-direct@shimadzu-biotech.jp.