Promega TNT™ T7 Insect Cell Extract Protein Expression System Companion Vectors
Designed for use with Insect Cell Extracts and contain untranslated region (UTR) sequences at the 5' and 3' ends of the gene coding region to enhance translation efficiency.
Supplier: Promega L1081
Description
- Protein Expression Vectors Used With Insect Cell Extracts for In Vitro Translation
- Efficient transfer of Flexi(R) vector inserts for direct use of recombinant clones, minimizing time wasted screening background colonies
- Adaptable to high-throughput formats for large screening projects
- Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or mammalian protein expression) as needed
Specifications
T7 | |
10mM Tris HCl, 1mM EDTA (pH 8) | |
100 ng/μL | |
pF25K ICE T7 Flexi™ vector |
SgfI and PmeI | |
-30°C to -10°C | |
20 μg | |
Clone ORF and conveniently transfer these inserts to a variety of vectors designed for multiple applications |
- Patent Pending.
- For research use only. Persons wishing to use this product or its derivatives in other fields of use, including without limitation, commercial sale, diagnostics or therapeutics, should contact Promega Corporation for licensing information.
- Use of this vector for applications outside of the TNT T7 Insect Cell Extract Protein Expression System may require a license from Shimadzu Corporation. For more information contact: t-direct@shimadzu-biotech.jp.