Promega Ultra Pure Deoxynucleotide Triphosphates (dNTPs) Sets DFS Item
High-quality dNTPs. Provided at a concentration of 100mM in water at pH 7.5.
Supplier:  Promega U1420
 View more versions of this product
Catalog No. PR-U1420
Explore available promotions
Description
Description
  • High-Quality dNTP Solutions for Your Workflow
  • Provided at 100mM in water at pH 7.5
Specifications
Specifications
7.5
100 mM
4 x 25 μmol
-30°C to -10°C
>99%
cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols