missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Wizard™ DNA Purification Resins
Used for purifying PCR-amplified double-stranded DNA.
$289.28 - $723.21
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | |||||
PR-A7141
![]() |
Promega
A7141 |
Minipreps Resin | 250 mL |
Each for $289.28
|
|
|||||
PR-A7181
![]() |
Promega
A7181 |
PCR Preps Resin | 250 mL |
Each for $355.89
|
|
|||||
PR-A7401
![]() |
Promega
A7401 |
Maxipreps Resin | 500 mL |
Each for $545.27
|
|
|||||
PR-A7701
![]() |
Promega
A7701 |
Midipreps Resin | 1000 mL |
Each for $723.21
|
|
|||||
Description
The resin is available with the systems and as a standalone product.
Specifications
15°C to 30°C | |
Liquid |