Promega Wizard™ DNA Purification Resins DFS Item
Used for purifying PCR-amplified double-stranded DNA.
Supplier:  Promega A7401
 View more versions of this product
Catalog No. PR-A7401
Explore available promotions
Description
Description
  • 300μg-1mg Plasmid DNA from 100-500mL Cultures
  • 3-hour protocol
  • Resin-based batch-binding and column-washing method
  • No organic extractions
Specifications
Specifications
15°C to 30°C
Purifies plasmid DNA
500 mL
Wizard™ Plus Maxipreps DNA Purification System
Maxipreps Resin
Liquid