Promega Wizard™ Plus SV Minipreps DNA Purification System
A silica membrane-based system for simple, rapid isolation of plasmid DNA from 1-10ml E. coli cultures. The miniprep procedure can be completed in 45 minutes or less.
Supplier: Promega A1460
Includes:
- DNA purification kit: Cell resuspension solution, cell lysis solution, neutralization solution, column wash solution, alkaline protease solution, nuclease-free water, miniprep spin columns, collection tubes with or without vacuum adapters
- DNA purification start-up kit (110V): Wizard™ Plus SV Minipreps DNA Purification System, Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold, vacuum adapters
Description
- 1-20μg High-Quality Plasmid DNA from 1-10mL Cultures
- Process up to 20 minipreps in 45 minutes
- Choice of spin or vacuum purification formats
- No phenol extractions or precipitations required
†Vacuum protocol requires use of a vacuum manifold.
Specifications
15°C to 30°C | |
Produces high yield plasmid DNA of consistent quality from bacterial cultures | |
250 Preps | |
1 to 20μg |
Spin or Vacuum | |
250 Wizard SV minicolumns, 250 Collection tubes, 100mL Column wash solution, 3mL Alkaline Protease solution, 100mL Neutralization solution, 75mL Cell resuspension solution, 75mL Cell Lysis solution, 25mL Nuclease-free water | |
DNA Purification System |
U.S. Patent 5,981,235, Australian Patent 729932.