missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Wizard™ SV 96 Genomic DNA Purification System
The Wizard™ SV 96 Genomic DNA Purification System provides a high-throughput, membrane-based technique for preparation of genomic DNA from cultured cells and tissue, including mouse tails.
$293.09 - $3445.35
Specifications
Content And Storage | 15°C to 30°C |
---|---|
For Use With (Application) | Provides fast, simple purification of intact DNA from tissue and cultured cells |
Includes: SV 96 Genomic DNA Purification Kit: 96-well deep-well plate, binding plate, 0.5M EDTA (pH 8.0), Nuclease-free water, Nuclei lysis solution, RNase A solution, Column wash solution, SV lysis buffer
SV 96 Genomic DNA Purification Start-Up Kit (110V): SV 96 Genomic DNA Purification System, Vac-Man™ 96 Vacuum Manifold, Welch™ Vacuum Pump for North America Electrical
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
PRA6780
|
Promega
A6780 |
SV 96 genomic DNA purification kit, 96-well deep-well plate, binding plate, 0.5M EDTA (pH 8), Nuclease-free Water, nuclei Lysis solution, RNase A solution, Column wash solution, SV Lysis Buffer |
Each for $3,445.35
|
|
|||||
PR-A2370
|
Promega
A2370 |
50mL Nuclei Lysis solution, 30mL pH 8 0.5M EDTA, 50mL Wizard™ SV Lysis Buffer, 185mL Wizard™ SV wash solution, 1mL RNase A solution, 150mL Nuclease-free Water, binding plate, 96-well deep well plate, DNA Elution plate, |
Each for $293.09
|
|
|||||
PR-A2371
|
Promega
A2371 |
2 x 50mL Nuclei Lysis solution, 30mL pH 8 0.5M EDTA, 3 x 50mL Wizard™ SV Lysis Buffer, 2 x 37mL Wizard™ SV wash solution, 2x 150mL Nuclease-free Water, 4 binding plate, 4 96-well deep well plate, protocol |
Each for $1,148.59
|
|
|||||
Description
- 96-Well, Vacuum-Based DNA Purification From Cells or Tissues
- Suitable for manual or high-throughput applications
- DNA from mouse tails in 45-60 minutes; DNA from cells in 30 minutes
- 20-30μg DNA per prep from 1.2cm mouse tail
Specifications
15°C to 30°C | |
Provides fast, simple purification of intact DNA from tissue and cultured cells |