Promega Wizard™ SV 96 Genomic DNA Purification System DFS Item
The Wizard™ SV 96 Genomic DNA Purification System provides a high-throughput, membrane-based technique for preparation of genomic DNA from cultured cells and tissue, including mouse tails.
Supplier:  Promega A2370
 View more versions of this product
Catalog No. PR-A2370
Explore available promotions
Includes:
  • SV 96 Genomic DNA Purification Kit: 96-well deep-well plate, binding plate, 0.5M EDTA (pH 8.0), Nuclease-free water, Nuclei lysis solution, RNase A solution, Column wash solution, SV lysis buffer
  • SV 96 Genomic DNA Purification Start-Up Kit (110V): SV 96 Genomic DNA Purification System, Vac-Man™ 96 Vacuum Manifold, Welch™ Vacuum Pump for North America Electrical
Requires : Manual, benchtop purification requires the Vac-Man™ 96 Vacuum Manifold (A2291) or a similar 96-well manifold; use with an automated liquid-handling workstation may require vacuum manifolds specific to the workstation.
Description
Description
  • 96-Well, Vacuum-Based DNA Purification From Cells or Tissues
  • Suitable for manual or high-throughput applications
  • DNA from mouse tails in 45-60 minutes; DNA from cells in 30 minutes
  • 20-30μg DNA per prep from 1.2cm mouse tail
Specifications
Specifications
15°C to 30°C
50mL Nuclei Lysis solution, 30mL pH 8 0.5M EDTA, 50mL Wizard™ SV Lysis Buffer, 185mL Wizard™ SV wash solution, 1mL RNase A solution, 150mL Nuclease-free Water, binding plate, 96-well deep well plate, DNA Elution plate,
System
Provides fast, simple purification of intact DNA from tissue and cultured cells
1 x 96 Preps