Promega Wizard™ SV 96 PCR Cleanup System DFS Item
The Wizard™ SV 96 PCR Clean-Up System is designed for high-throughput purification of 100bp-10kb PCR products from excess nucleotides, primers and primer dimers.
Supplier:  Promega A9342
 View more versions of this product
Catalog No. PR-A9342
Explore available promotions
Includes:
  • PCR Clean-Up kit: Membrane binding solution, Nuclease-free water, Binding plate, Collection plate (except A9340)
  • PCR Clean-Up start-up kit (110V): Wizard™ SV 96 PCR Clean-Up System, Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold
Requires : Requires the Vac-Man 96 Vacuum Manifold for manual protocols; automated protocols require a vacuum manifold compatible with the automated system
Description
Description
  • 96-Well, Vacuum-Based Purification of PCR DNA
  • Protocol complete in as little as 20 minutes
  • Purifies 100bp to 10kb PCR Products
  • 96-well filter plate processing
Specifications
Specifications
15°C to 30°C
High-throughput purification of 100bp to 10kb PCR products from excess Nucleotides, primers and primer dimers
8 x 96 Preps
96-well Plate
200mL Membrane binding solution, 300mL Nuclease-free Water, 8 binding plates, 8 collection plate, 1 protocol
System