Promega Wizard™ SV Gel and PCR Cleanup System DFS Item
The Wizard™ SV Gel and PCR Clean-Up System is designed to purify DNA fragments of 100bp-10kb from standard or low-melting agarose gels or to purify products directly from PCR or restriction digests.
Supplier:  Promega A9281
 View more versions of this product
Catalog No. PR-A9281
Explore available promotions
Description
Description
  • Purifies DNA Fragments from Gels or PCR in 15 Minutes
  • Choice of spin- or vacuum-based protocols
  • Purifies 100bp to 10kb DNA fragments
Specifications
Specifications
15°C to 30°C
Purifies DNA from agarose gels or PCR reactions in as little as 15 min.
50 Preps
Spin or Vacuum
20mL Membrane binding solution, 15mL membrane wash solution, 3.75mL Nuclease-free Water, 50 Wizard™ SV minicolumns, 50 2mL collection tubes
System