Learn More
Propargyl acetate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 380820050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Propargyl acetate | |
0.9890g/mL | |
33°C | |
96% min. (GC) | |
C5H6O2 | |
CH≡CCH2OCOCH3 | |
5g | |
propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference | |
CC(=O)OCC#C | |
98.1 | |
98.1 |
627-09-8 | |
27.0°C to 28.0°C (8.0mmHg) | |
Authentic | |
Glass bottle | |
1.4160 to 1.4180 | |
MFCD00041601 | |
0.989 | |
RIZZXCJMFIGMON-UHFFFAOYSA-N | |
prop-2-ynyl acetate | |
69388 | |
97% |
Chemical Identifiers
627-09-8 | |
98.1 | |
RIZZXCJMFIGMON-UHFFFAOYSA-N | |
69388 | |
CC(=O)OCC#C |
C5H6O2 | |
MFCD00041601 | |
propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference | |
prop-2-ynyl acetate |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin
GHS Signal Word: Danger
RUO â Research Use Only