missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Propargyl acetate, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 627-09-8 |
---|---|
Molecular Formula | C5H6O2 |
Molecular Weight (g/mol) | 98.1 |
MDL Number | MFCD00041601 |
InChI Key | RIZZXCJMFIGMON-UHFFFAOYSA-N |
Synonym | propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference |
PubChem CID | 69388 |
IUPAC Name | prop-2-ynyl acetate |
SMILES | CC(=O)OCC#C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC380820050
|
Thermo Scientific Chemicals
380820050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC380820250
|
Thermo Scientific Chemicals
380820250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
627-09-8 | |
98.1 | |
RIZZXCJMFIGMON-UHFFFAOYSA-N | |
69388 | |
CC(=O)OCC#C |
C5H6O2 | |
MFCD00041601 | |
propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference | |
prop-2-ynyl acetate |
Specifications
627-09-8 | |
27.0°C to 28.0°C (8.0mmHg) | |
Authentic | |
Glass bottle | |
1.4160 to 1.4180 | |
MFCD00041601 | |
0.989 | |
RIZZXCJMFIGMON-UHFFFAOYSA-N | |
prop-2-ynyl acetate | |
69388 | |
97% |
0.9890g/mL | |
33°C | |
96% min. (GC) | |
C5H6O2 | |
CH≡CCH2OCOCH3 | |
5g | |
propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference | |
CC(=O)OCC#C | |
98.1 | |
98.1 | |
Propargyl acetate |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin
GHS Signal Word: Danger
RUO â Research Use Only