missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propargyl acetate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 380820250

 View more versions of this product

Catalog No. AC380820250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Propargyl acetate
0.9890g/mL
33°C
96% min. (GC)
C5H6O2
CH≡CCH2OCOCH3
25g
propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference
CC(=O)OCC#C
98.1
98.1
627-09-8
27.0°C to 28.0°C (8.0mmHg)
Authentic
Glass bottle
1.4160 to 1.4180
MFCD00041601
0.989
RIZZXCJMFIGMON-UHFFFAOYSA-N
prop-2-ynyl acetate
69388
97%

Chemical Identifiers

627-09-8
98.1
RIZZXCJMFIGMON-UHFFFAOYSA-N
69388
CC(=O)OCC#C
C5H6O2
MFCD00041601
propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference
prop-2-ynyl acetate
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only