missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propionic acid, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 149300010

 View more versions of this product

Catalog No. AC149300010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Propionic acid
0.9900g/mL
99%
C3H6O2
C2H5CO2H
02, 234
15, 7939
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propanoic acid
1032
74.08
≥98.5% (GC)
Liquid
79-09-4
Authentic
EcoSafPak™ Glass Bottle
1.3850 to 1.3870 (20°C, 589nm)
1 L
12,415
propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
CCC(=O)O
74.079
CHEBI:30768
5mbar at 20°C
Pure

Chemical Identifiers

79-09-4
propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
CHEBI:30768
CCC(=O)O
XBDQKXXYIPTUBI-UHFFFAOYSA-N
1032
propanoic acid
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only