Learn More
Propionic acid, 99%, pure, Thermo Scientific Chemicals
$83.81 - $375.31
Chemical Identifiers
CAS | 79-09-4 |
---|---|
InChI Key | XBDQKXXYIPTUBI-UHFFFAOYSA-N |
Synonym | propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin |
PubChem CID | 1032 |
ChEBI | CHEBI:30768 |
IUPAC Name | propanoic acid |
SMILES | CCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC149300010
|
Thermo Scientific Chemicals
149300010 |
1 L | EcoSafPak™ Glass Bottle |
Each for $83.81
|
|
|||||
AC149300025
|
Thermo Scientific Chemicals
149300025 |
2.5 L | Glass Bottle |
Each for $180.70
Case of 4 Each for $0.00
|
|
|||||
AC149300100
|
Thermo Scientific Chemicals
149300100 |
10 L | Plastic drum |
Each for $375.31
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-09-4 | |
propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin | |
CHEBI:30768 | |
CCC(=O)O |
XBDQKXXYIPTUBI-UHFFFAOYSA-N | |
1032 | |
propanoic acid |
Specifications
79-09-4 | |
Authentic | |
EcoSafPak™ Glass Bottle | |
1.3850 to 1.3870 (20°C, 589nm) | |
1 L | |
12,415 | |
propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin | |
CCC(=O)O | |
74.079 | |
CHEBI:30768 | |
5mbar at 20°C | |
Pure | |
Propionic acid |
0.9900g/mL | |
99% | |
C3H6O2 | |
C2H5CO2H | |
02, 234 | |
15, 7939 | |
XBDQKXXYIPTUBI-UHFFFAOYSA-N | |
propanoic acid | |
1032 | |
74.08 | |
≥98.5% (GC) | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
May be corrosive to metals.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 201-176-3
RUO – Research Use Only