missing translation for 'onlineSavingsMsg'
Learn More

Propionic acid, 99%, pure

CAS: 79-09-4 | C3H6O2 | 74.079 g/mol

Supplier:  Thermo Scientific Chemicals 149300025

 View more versions of this product

Catalog No. AC149300025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Propionic acid
-22.0°C
141.0°C
Authentic
Glass Bottle
1.3850 to 1.3870
2.5 L
12,415
15, 7939
Solubility in water: soluble.
CCC(=O)O
74.079
CHEBI:30768
5mbar at 20°C
Pure
79-09-4
0.9900g/mL
51°C
99%
C3H6O2
C2H5CO2H
02, 234
0.99
propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propanoic acid
1032
74.08
99%
Liquid

Chemical Identifiers

79-09-4
propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
CHEBI:30768
CCC(=O)O
XBDQKXXYIPTUBI-UHFFFAOYSA-N
1032
propanoic acid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only