missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propionic acid, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 149300100

 View more versions of this product

Catalog No. AC149300100

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Propionic acid
-22.0°C
141.0°C
Authentic
Plastic drum
1.3850 to 1.3870
10 L
12,415
15, 7939
Solubility in water: soluble.
CCC(=O)O
74.08
1.02 mPa.s (25°C)
74.08
99%
Liquid
79-09-4
0.9900g/mL
51°C
98.5% min. (GC)
C3H6O2
C2H5CO2H
02, 234
0.99
propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propanoic acid
1032
CHEBI:30768
5mbar at 20°C
Pure

Chemical Identifiers

79-09-4
propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
CHEBI:30768
CCC(=O)O
XBDQKXXYIPTUBI-UHFFFAOYSA-N
1032
propanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
May be corrosive to metals.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 201-176-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only