Propper Chex-All™ Heat Sealable Sterilization Pouches
Heat-Sealable Chex-All™ Sterilization Pouches are premium quality sterile packaging products designed to reduce sterilization processing costs. For use in steam and ethylene oxide. Pre-printed indicator inside and outside the pouch. Triple-sealed paper/plastic film ensures content security.
Supplier:  Propper 02501400
 View more versions of this product
Catalog No. 15-234-18C
Explore available promotions
Description
Description
Heat-sealable
Chex-All™ Heat Seal Sterilization Pouches provide a sterile barrier, and feature easy-to-read pre-printed indicators (both inside and outside the pouch). Heat-Sealing pouches provide the convenience of custom sizing to meet the individual needs of your instruments.
  • Chex-All™ pouches are for use in steam and ethylene oxide, eliminating the need for two sets of pouches.
  • Chex-All™ pouches are validated for double pouch packaging.
  • Heat Seal Sterilization Pouches allow you to choose the exact length your need.
  • Easy-to-read chemical indicator changes from yellow to black in steam.
  • Easy-to-read chemical indicator changes from red to green in EO.
  • Made of 2.5 mm plastic and 73-74 gram.
  • Triple sealed paper/plastic film provides excellent sterility protection.
Specifications
Specifications
Autoclavable
White
Indicator imprints on the inside and outside for both steam and ethylene oxide gas
Non-sterile
500/Cs.
16 x 8 in. (40.6 x 20.3 cm)
40.6 cm
20.3 cm
Heat Seal
Single-use
Paper, Plastic
0.064 mm
2.50 mil
16 in.
8 in.