Learn More
(±)-Propylene oxide, 99%, pure
(+/-)-Propylene oxide, 99%, C3H6O, CAS Number-75-56-9, 2,3-epoxypropane, epoxypropane, methyloxirane, propylene oxide, oxirane, methyl, propyleneoxide, 1,2-propylene oxide, propene oxide, propylene epoxide, 1,2-epoxypropane, 1L, 17, 6, 34.0 deg.C, CHEBI:38685, Undesignated, 0.8300g/mL, 200-879-2 | CAS: 75-56-9 | C3H6O | 58.08 g/mol
Supplier: Thermo Scientific Chemicals 149620010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (±)-Propylene oxide | |
| 75-56-9 | |
| 0.8300g/mL | |
| −37°C | |
| 99% | |
| C3H6O | |
| MFCD00005126 | |
| 17, 6 | |
| 15, 7969 | |
| GOOHAUXETOMSMM-UHFFFAOYNA-N | |
| 2-methyloxirane | |
| 6378 | |
| 58.08 | |
| 99% | |
| Liquid |
| 99% | |
| -112.0°C | |
| 34.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3650 to 1.3670 | |
| 1 L | |
| 0.83 | |
| propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane | |
| CC1CO1 | |
| 58.08 | |
| CHEBI:38685 | |
| 590mbar at 20°C | |
| Pure |
Chemical Identifiers
| 75-56-9 | |
| 58.08 | |
| GOOHAUXETOMSMM-UHFFFAOYNA-N | |
| 6378 | |
| 2-methyloxirane |
| C3H6O | |
| MFCD00005126 | |
| propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane | |
| CHEBI:38685 | |
| CC1CO1 |
Safety and Handling
GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.
May cause genetic defects.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wash face, hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Danger
EINECSNumber : 200-879-2
RTECSNumber : TZ2975000
TSCA : TSCA
RUO – Research Use Only