missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-Propylene oxide, 99%, pure, Thermo Scientific Chemicals

(+/-)-Propylene oxide, 99%, C3H6O, CAS Number-75-56-9, 2,3-epoxypropane, epoxypropane, methyloxirane, propylene oxide, oxirane, methyl, propyleneoxide, 1,2-propylene oxide, propene oxide, propylene epoxide, 1,2-epoxypropane, 1L, 17, 6, 34.0 deg.C, CHEBI:38685, Undesignated, 0.8300g/mL, 200-879-2

$115.42 - $186.22

Chemical Identifiers

CAS 75-56-9
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005126
InChI Key GOOHAUXETOMSMM-UHFFFAOYNA-N
Synonym propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
PubChem CID 6378
ChEBI CHEBI:38685
IUPAC Name 2-methyloxirane
SMILES CC1CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC149620010
SDS View Documents
Thermo Scientific Chemicals
149620010
1 L Glass bottle
Each for $115.42
Case of 6 Each for $0.00
 
AC149620025
SDS View Documents
Thermo Scientific Chemicals
149620025
2.5 L Glass bottle
Each for $186.22
Case of 4 Each for $0.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-56-9
58.08
GOOHAUXETOMSMM-UHFFFAOYNA-N
6378
2-methyloxirane
C3H6O
MFCD00005126
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
CHEBI:38685
CC1CO1

Specifications

75-56-9
0.8300g/mL
−37°C
99%
C3H6O
MFCD00005126
17, 6
15, 7969
GOOHAUXETOMSMM-UHFFFAOYNA-N
2-methyloxirane
6378
58.08
99%
Liquid
-112.0°C
34.0°C
Authentic
Glass bottle
1.3650 to 1.3670
1 L
0.83
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
CC1CO1
58.08
CHEBI:38685
590mbar at 20°C
Pure
(±)-Propylene oxide, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 200-879-2

RTECSNumber : TZ2975000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only