Contec™ PROSAT™ Presaturated Wipes in SatBox™ Containers
Reduce solvent usage with presatured wipes. Contec™ PROSAT™ Presaturated Wipes in SatBox™ Containers are ideal for wiping process tools and other equipment, as well as many other cleaning applications in critical environments. The reusable SatBox container is made from high-density polypropylene and reduces solvent evaporation.
Supplier:  Contec™ CSCN0002
 View more versions of this product
Catalog No. 19-084-300
Explore available promotions
Description
Description
  • Premoistened knitted polyester wipers and nonwoven polypropylene wipes are saturated with various solvent blends
  • Polynit Heatseal wipers are standard weight, textured 100% polyester interlock knit with nonabrasive, heatsealed edges
  • Polyester wipes are abrasion and chemical resistant
  • Economical nonwoven wipes are made of 100% meltblown polypropylene
  • Ideal for wiping process tools and other equipment, as well as many other cleaning applications in critical environments
  • Reusable SatBox container is made from high-density polypropylene and is compatible with various blends of isopropyl alcohol, ethanol, and acetone
  • SatBox reduces solvent evaporation.
Specifications
Specifications
SatBox Container
24 cm
24.13 cm
9.5 x 6.25 in. (25 x 17 cm)
9.5 in.
9.5 in.
HDPE
Controlled environment cleaning