Contec™ PROSAT™ Presaturated Wipes in SatBox™ Containers
Reduce solvent usage with presatured wipes. Contec™ PROSAT™ Presaturated Wipes in SatBox™ Containers are ideal for wiping process tools and other equipment, as well as many other cleaning applications in critical environments. The reusable SatBox container is made from high-density polypropylene and reduces solvent evaporation.
$63.10 - $63.10
Catalog Number | Mfr. No. | Product Type | Dimensions (L x W) | Material | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Dimensions (L x W) | Material | Price | Quantity | |||
19-084-300
|
Contec™
CSCN0002 |
SatBox Container | 9.5 x 6.25 in. (25 x 17 cm) | HDPE |
Case of 4 Each for $63.10
|
|
|||
Description
- Premoistened knitted polyester wipers and nonwoven polypropylene wipes are saturated with various solvent blends
- Polynit Heatseal wipers are standard weight, textured 100% polyester interlock knit with nonabrasive, heatsealed edges
- Polyester wipes are abrasion and chemical resistant
- Economical nonwoven wipes are made of 100% meltblown polypropylene
- Ideal for wiping process tools and other equipment, as well as many other cleaning applications in critical environments
- Reusable SatBox container is made from high-density polypropylene and is compatible with various blends of isopropyl alcohol, ethanol, and acetone
- SatBox reduces solvent evaporation.